Stampa…

Anti-TEAD2 Mouse Monoclonal Antibody [clone: 8C324]

Fornitore: United States Biological
USBIT2140-20EA 547 CHF
USBIT2140-20
Anti-TEAD2 Mouse Monoclonal Antibody [clone: 8C324]
Anticorpi
Recommended dilution: Immunochemistry: 2-100 ?g/ml

Type: Primary
Antigen: TEAD2 (TEA domain family member 2)
Clonality: monoclonal
Clone: 8C324
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR