Stampa…

Anti-TPST2 Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT2040-37A-BIOTEA 1200 CHF
USBIT2040-37A-BIOT
Anti-TPST2 Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: TPST2 (tyrosylprotein sulfotransferase 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR