Stampa…

Anti-TCR, gamma, delta Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K24]

Fornitore: United States Biological
USBIT2040-06GEA 632 CHF
USBIT2040-06G
Anti-TCR, gamma, delta Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K24]
Anticorpi
Type: Primary
Antigen: TCR, gamma, delta
Clonality: monoclonal
Clone: 5K24
Conjugation: FITC (Fluorescein)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR