Stampa…

Anti-CD3D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K51]

Fornitore: United States Biological
USBIT2040-05MEA 1030 CHF
USBIT2040-05M
Anti-CD3D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K51]
Anticorpi
Do not freeze!

Type: Primary
Antigen: CD3D (CD3d molecule, delta (CD3-TCR complex))
Clonality: monoclonal
Clone: 10K51
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR