Stampa…

Anti-TCR DO11.10 Mouse Monoclonal Antibody

Fornitore: United States Biological
USBIT2040-04VEA 406 CHF
USBIT2040-04V
Anti-TCR DO11.10 Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: TCR DO11.10
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR