Stampa…

Anti-TCR, alpha, beta Mouse Monoclonal Antibody

Fornitore: United States Biological
USBIT2040-03CEA 540 CHF
USBIT2040-03C
Anti-TCR, alpha, beta Mouse Monoclonal Antibody
Anticorpi
Type: Primary
Antigen: TCR, alpha, beta
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Monkey, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR