Stampa…

Anti-TCR, alpha, beta, V beta 2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5K83]

Fornitore: United States Biological
USBIT2040-02W4EA 655 CHF
USBIT2040-02W4
Anti-TCR, alpha, beta, V beta 2 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5K83]
Anticorpi
Type: Primary
Antigen: TCR, alpha, beta, V beta 2
Clonality: monoclonal
Clone: 5K83
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR