Stampa…

Anti-T-complex Protein 1 Subunit gamma Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT2035-11-BIOTEA 1200 CHF
USBIT2035-11-BIOT
Anti-T-complex Protein 1 Subunit gamma Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: T-complex Protein 1 Subunit gamma
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR