Stampa…

Anti-TCF4 Rabbit Monoclonal Antibody [clone: 7H75]

Fornitore: United States Biological
USBIT2026-03EA 739 CHF
USBIT2026-03
Anti-TCF4 Rabbit Monoclonal Antibody [clone: 7H75]
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50; ChImmunoprecipitation: 1:50

Type: Primary
Antigen: TCF4 (transcription factor 4)
Clonality: monoclonal
Clone: 7H75
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR