Stampa…

Anti-HNF1A Rabbit Polyclonal Antibody (Biotin)

Fornitore: United States Biological
USBIT2020-22-BIOTEA 1200 CHF
USBIT2020-22-BIOT
Anti-HNF1A Rabbit Polyclonal Antibody (Biotin)
Anticorpi
Type: Primary
Antigen: HNF1A (HNF1 homeobox A)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR