Stampa…

Anti-HNF1A Rabbit Monoclonal Antibody [clone: 7H74]

Fornitore: United States Biological
USBIT2020-03EA 753 CHF
USBIT2020-03
Anti-HNF1A Rabbit Monoclonal Antibody [clone: 7H74]
Anticorpi
Recommended dilution: Flow Cytometry: 1:100; Immunofluorescence: 1:50; Immunochemistry: 1:100; Immunoprecipitation: 1:50; Western Blot: 1:1000

Type: Primary
Antigen: HNF1A (HNF1 homeobox A)
Clonality: monoclonal
Clone: 7H74
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR