Stampa…

Anti-T Cell Lymphoma 1A Chicken Polyclonal Antibody

Fornitore: United States Biological
USBIT2017-10EA 667 CHF
USBIT2017-10
Anti-T Cell Lymphoma 1A Chicken Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: T Cell Lymphoma 1A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR