Stampa…

Anti-IL27RA Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT2009-04AEA 770 CHF
USBIT2009-04A
Anti-IL27RA Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-1.0 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: IL27RA (interleukin 27 receptor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR