Stampa…

Anti-TBX15 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT1848-38EA 889 CHF
USBIT1848-38
Anti-TBX15 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:1000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: TBX15 (T-box 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR