Stampa…

Anti-TBX3 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT1848-24BEA 1100 CHF
USBIT1848-24B
Anti-TBX3 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TBX3 (T-box 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR