Stampa…

Anti-TBX1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT1848-24EA 924 CHF
USBIT1848-24
Anti-TBX1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunoprecipitation: 1-3?g/ml; Western Blot: 1-3?g/ml

Type: Primary
Antigen: TBX1 (T-box 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR