Stampa…

Anti-TBL1X Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT1845-01EA 903 CHF
USBIT1845-01
Anti-TBL1X Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TBL1X (transducin (beta)-like 1X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR