Stampa…

Anti-MAPT Mouse Monoclonal Antibody [clone: 6A193]

Fornitore: United States Biological
USBIT1040-02DEA 917 CHF
USBIT1040-02D
Anti-MAPT Mouse Monoclonal Antibody [clone: 6A193]
Anticorpi
Recommended dilution: ELISA: 3-5 ?g/ml; Western Blot: 1-3?g/ml

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: 6A193
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR