Stampa…

Anti-MAPT Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT1032-74EA 836 CHF
USBIT1032-74
Anti-MAPT Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR