Stampa…

Anti-MAPT Mouse Monoclonal Antibody [clone: 7H166]

Fornitore: United States Biological
USBIT1030-04EA 1050 CHF
USBIT1030-04
Anti-MAPT Mouse Monoclonal Antibody [clone: 7H166]
Anticorpi
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 1-2?g/ml

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: 7H166
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR