Stampa…

Anti-MAPT Mouse Monoclonal Antibody [clone: 5H68]

Fornitore: United States Biological
USBIT1030-03EA 945 CHF
USBIT1030-03
Anti-MAPT Mouse Monoclonal Antibody [clone: 5H68]
Anticorpi
Recommended dilution: Immunoblot: A 1:500-1:2000

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: 5H68
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR