Stampa…

Anti-TASP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Fornitore: United States Biological
USBIT1023-90A-FITCEA 1200 CHF
USBIT1023-90A-FITC
Anti-TASP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anticorpi
Recommended dilution: FLISA: 1:1000; Western Blot: 1:250-1:500; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: TASP1 (taspase, threonine aspartase, 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR