Stampa…

Anti-Tankyrase Mouse Monoclonal Antibody [clone: 11C843]

Fornitore: United States Biological
USBIT0900-20BEA 929 CHF
USBIT0900-20B
Anti-Tankyrase Mouse Monoclonal Antibody [clone: 11C843]
Anticorpi
Recommended dilution: ELISA: 1 ?g/ml-3ng/ml; Western Blot: 1:500-1:1000; Immunochemistry: 1. 5 ?g/ml

Type: Primary
Antigen: Tankyrase
Clonality: monoclonal
Clone: 11C843
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR