Stampa…

Anti-Taleranol Sheep Polyclonal Antibody

Fornitore: United States Biological
USBIT0730EA 1080 CHF
USBIT0730
Anti-Taleranol Sheep Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 0. 3125 ?g/ml

Type: Primary
Antigen: Taleranol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR