Stampa…
Anti-TACR2 Rabbit Polyclonal Antibody
Fornitore: United States Biological
USBIT0501-20BEA 1150 CHF
USBIT0501-20B
Anti-TACR2 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 40 ?g/ml
Type: Primary
Antigen: TACR2 (tachykinin receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Mehr About VWR