Stampa…

Anti-ADAM17 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT0500-01LEA 681 CHF
USBIT0500-01L
Anti-ADAM17 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: ADAM17 (ADAM metallopeptidase domain 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR