Stampa…

Anti-TNFRSF13B Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT0294-01NEA 889 CHF
USBIT0294-01N
Anti-TNFRSF13B Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 5 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: TNFRSF13B (tumor necrosis factor receptor superfamily, member 13B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR