Stampa…

Anti-TNKS Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT0292-03EA 774 CHF
USBIT0292-03
Anti-TNKS Goat Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-1. 5 ?g/ml

Type: Primary
Antigen: TNKS (tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR