Stampa…

Anti-TAB3 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIT0291-91EA 756 CHF
USBIT0291-91
Anti-TAB3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: TAB3 (TGF-beta activated kinase 1/MAP3K7 binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR