Stampa…

Anti-TAB1 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIT0291-86EA 1090 CHF
USBIT0291-86
Anti-TAB1 Goat Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: TAB1 (TGF-beta activated kinase 1/MAP3K7 binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR