Stampa…

Anti-Gamma- Synuclein Rabbit polyclonal antibody

Fornitore: United States Biological
USBIS9502-18EA 1110 CHF
USBIS9502-18
Anti-Gamma- Synuclein Rabbit polyclonal antibody
Anticorpi
Type: Primary
Antigen: Gamma- Synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR