Stampa…

Anti-SNCB Sheep Polyclonal Antibody

Fornitore: United States Biological
USBIS9501-02GEA 761 CHF
USBIS9501-02G
Anti-SNCB Sheep Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: SNCB (synuclein, beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR