Stampa…

Anti-Synuclein, alpha Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIS9500-27AEA 642 CHF
USBIS9500-27A
Anti-Synuclein, alpha Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: Synuclein, alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR