Stampa…

Anti-Synuclein, alpha Rabbit Monoclonal Antibody [clone: 10E50]

Fornitore: United States Biological
USBIS9500-21CEA 776 CHF
USBIS9500-21C
Anti-Synuclein, alpha Rabbit Monoclonal Antibody [clone: 10E50]
Anticorpi
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50; Immunochemistry: 1:200

Type: Primary
Antigen: Synuclein, alpha
Clonality: Monoclonal
Clone: 1E+51
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR