Stampa…

Anti-STXBP1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIS9205-80EA 788 CHF
USBIS9205-80
Anti-STXBP1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: STXBP1 (syntaxin binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR