Stampa…

Anti-STX6 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIS9205-25EEA 774 CHF
USBIS9205-25E
Anti-STX6 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 3-1 ?g/ml

Type: Primary
Antigen: STX6 (syntaxin 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR