Stampa…

Anti-SNCAIP Goat Polyclonal Antibody

Fornitore: United States Biological
USBIS9116-05AEA 860 CHF
USBIS9116-05A
Anti-SNCAIP Goat Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 1:400; ELISA: 1:5000-1:10000; Western Blot: 1:2000

Type: Primary
Antigen: SNCAIP (synuclein, alpha interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR