Stampa…

Anti-SDC2 Rat Monoclonal Antibody [clone: 7H78]

Fornitore: United States Biological
USBIS9111-40B1EA 1070 CHF
USBIS9111-40B1
Anti-SDC2 Rat Monoclonal Antibody [clone: 7H78]
Anticorpi
Recommended dilution: ELISA: 0.5-1 ?g/ml; Immunochemistry: 25 ?g/ml

Type: Primary
Antigen: SDC2 (syndecan 2)
Clonality: Monoclonal
Clone: 7H78
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR