Stampa…

Anti-SDC1 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIS9111-30JEA 889 CHF
USBIS9111-30J
Anti-SDC1 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunochemistry: 15 ?g/ml

Type: Primary
Antigen: SDC1 (syndecan 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR