Stampa…

Anti-Synaptic adhesion-like molecule 2 Mouse Monoclonal Antibody [clone: 11n180]

Fornitore: United States Biological
USBIS9111-04AEA 1030 CHF
USBIS9111-04A
Anti-Synaptic adhesion-like molecule 2 Mouse Monoclonal Antibody [clone: 11n180]
Anticorpi
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: Synaptic adhesion-like molecule 2
Clonality: Monoclonal
Clone: 11n180
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR