Stampa…

Anti-SYT4 Rabbit Polyclonal Antibody (Cy5®)

Fornitore: United States Biological
USBIS9109-40EEA 1450 CHF
USBIS9109-40E
Anti-SYT4 Rabbit Polyclonal Antibody (Cy5®)
Anticorpi
Type: Primary
Antigen: SYT4 (synaptotagmin IV)
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR