Stampa…

Anti-SYT1 Goat Polyclonal Antibody

Fornitore: United States Biological
USBIS9109-19HEA 889 CHF
USBIS9109-19H
Anti-SYT1 Goat Polyclonal Antibody
Anticorpi
Recommended dilution: ELISA: 1:2000; Western Blot: 0. 3-1 ?g/ml; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: SYT1 (synaptotagmin I)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR