Stampa…

Anti-Synaptotagmin Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIS9108-99CEA 533 CHF
USBIS9108-99C
Anti-Synaptotagmin Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Immunofluorescence:1:100-1:200

Type: Primary
Antigen: Synaptotagmin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR