Stampa…

Anti-SYP Rabbit Monoclonal Antibody [clone: 11C1075]

Fornitore: United States Biological
USBIS9108-20AEA 784 CHF
USBIS9108-20A
Anti-SYP Rabbit Monoclonal Antibody [clone: 11C1075]
Anticorpi
Recommended dilution: Immunofluorescence: 1:400; Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: SYP (synaptophysin)
Clonality: Monoclonal
Clone: 11C1075
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR