Stampa…

Anti-SYP Mouse Monoclonal Antibody [clone: 5F304]

Fornitore: United States Biological
USBIS9108-09BEA 871 CHF
USBIS9108-09B
Anti-SYP Mouse Monoclonal Antibody [clone: 5F304]
Anticorpi
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: SYP (synaptophysin)
Clonality: Monoclonal
Clone: 5F304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR