Stampa…

Anti-SYNGR3 Rabbit Polyclonal Antibody

Fornitore: United States Biological
USBIS9106-52AEA 1060 CHF
USBIS9106-52A
Anti-SYNGR3 Rabbit Polyclonal Antibody
Anticorpi
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: SYNGR3 (synaptogyrin 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR