Imprimer

D(+)-Ribono-1,4-lactone 96%

Fournisseur: Thermo Scientific

Synonyms: Acide D(+)-Ribonique γ-lactone

ACRO227830100EA 302 CHF
ACRO227830100
D(+)-Ribono-1,4-lactone 96%
D(+)-Ribono-1,4-lactone
Formule: C₅H₈O₅
MW: 148,11 g/mol
Melting Pt: 83…85 °C
température de stockage: Réfrigérateur
MDL Number: MFCD00063241
Numéro CAS: 5336-08-3

Order Now

Résultats des tests de spécification

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
GC ≥95 %
Residue after ignition ≤0.2 %
Specific optical rotation +17° to +19° (20°C, 589 nm) (c=5, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR