Imprimer

Acide L(+)-tartrique ≥99% ACS

Danger

J316-100GEA 127 CHF
J316-100G
Acide L(+)-tartrique ≥99% ACS
Acide L(+)-tartrique

Chelates Na+, K+ monovalent cations.

Formule: C₄H₆O₆
MW: 150,09 g/mol
Point d'ébullition: 399 °C (1013 hPa)
Melting Pt: 168…170 °C
Densité: 1,759 g/cm³ (20 °C)
Point d'éclair: 150 °C (coupe fermée)
MDL Number: MFCD00064207
Numéro CAS: 87-69-4
Merck Index: 12,09237
https://ch.vwr.com/stibo/web/std.lang.all/72/32/2297232.jpg

Order Now

Résultats des tests de spécification

Chloride 0.001%
Heavy Metals (as Pb) ≤5 ppm
Insolubles ≤0.005%
Iron ≤5 ppm
Oxalate Test Pass
Phosphate ≤0.001%
Purity ≥99%
Residue after Ignition ≤0.02%
Sulphur Compounds ≤0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR