Nα-Acétyl-DL-tryptophane
Synonyms:
Ac-DL-Trp-OH
, Nα-Acétyl-DL-tryptophane
EDQMA0208000EA
199
CHF
EDQMA0208000
Nα-Acétyl-DL-tryptophane
N-Acétyl-DL-tryptophane
Formule:
C₁₃H₁₄N₂O₃ MW: 246,27 g/mol Melting Pt: 205…206 °C |
MDL Number:
MFCD00005644 Numéro CAS: 87-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...