Imprimer
Cryostat, CM1850
Fournisseur: Leica
720-1884EA 0 CHF
LEIC1491850UVEL
Cryostat, CM1850
Up-to-date technology with an ergonomic design for quick and safe sectioning with convenient foot rest. The chamber temperature goes down to −35 °C. Actively cooled quick freeze shelf with Peltier unit for freezing down to −60 °C.
  • Power-saving refrigeration system
  • Splash-protected microtome
  • Motorised coarse feed at two speeds
  • Smooth specimen orientation with automatic centering
  • Automatic and manual chamber defrost cycles
A versatile cryostat with optimised cooling system, rapid specimen freezing and smooth specimen orientation for the high-quality sectioning demanded in routine histology and clinical pathology.